MilliporeSigma
  • Progesterone receptor membrane component 1 inhibits tumor necrosis factor alpha induction of gene expression in neural cells.
Progesterone receptor membrane component 1 inhibits tumor necrosis factor alpha induction of gene expression in neural cells.
PloS one (2019-04-27)
Karlie A Intlekofer, Kelsey Clements, Haley Woods, Hillary Adams, Alexander Suvorov, Sandra L Petersen
ABSTRACT
Progesterone membrane receptor component 1 (Pgrmc1) is a cytochrome b5-related protein with wide-ranging functions studied most extensively in non-neural tissues. We previously demonstrated that Pgrmc1 is widely distributed in the brain with highest expression in the limbic system. To determine Pgrmc1 functions in cells of these regions, we compared transcriptomes of control siRNA-treated and Pgrmc1 siRNA-treated N42 hypothalamic cells using whole genome microarrays. Our bioinformatics analyses suggested that Pgrmc1 plays a role in immune functions and likely regulates proinflammatory cytokine signaling. In follow-up studies, we showed that one of these cytokines, TNFα, increased expression of rtp4, ifit3 and gbp4, genes found on microarrays to be among the most highly upregulated by Pgrmc1 depletion. Moreover, either Pgrmc1 depletion or treatment with the Pgrmc1 antagonist, AG-205, increased both basal and TNFα-induced expression of these genes in N42 cells. TNFα had no effect on levels of Rtp4, Ifit3 or Gbp4 mRNAs in mHippoE-18 hippocampal control cells, but Pgrmc1 knock-down dramatically increased basal and TNFα-stimulated expression of these genes. P4 had no effect on gbp4, ifit3 or rtp4 expression or on the ability of Pgrmc1 to inhibit TNFα induction of these genes. However, a majority of the top upstream regulators of Pgrmc1 target genes were related to synthesis or activity of steroids, including P4, that exert neuroprotective effects. In addition, one of the identified Pgrmc1 targets was Nr4a1, an orphan receptor important for the synthesis of most steroidogenic molecules. Our findings indicate that Pgrmc1 may exert neuroprotective effects by suppressing TNFα-induced neuroinflammation and by regulating neurosteroid synthesis.