MilliporeSigma
Studies on transformation of Escherichia coli with plasmids.
Journal of molecular biology (1983-06-05)
D Hanahan
ABSTRACT
Factors that affect the probability of genetic transformation of Escherichia coli by plasmids have been evaluated. A set of conditions is described under which about one in every 400 plasmid molecules produces a transformed cell. These conditions include cell growth in medium containing elevated levels of Mg2+, and incubation of the cells at 0 degrees C in a solution of Mn2+, Ca2+, Rb+ or K+, dimethyl sulfoxide, dithiothreitol, and hexamine cobalt (III). Transformation efficiency declines linearly with increasing plasmid size. Relaxed and supercoiled plasmids transform with similar probabilities. Non-transforming DNAs compete consistent with mass. No significant variation is observed between competing DNAs of different source, complexity, length or form. Competition with both transforming and non-transforming plasmids indicates that each cell is capable of taking up many DNA molecules, and that the establishment of a transformation event is neither helped nor hindered significantly by the presence of multiple plasmids.
Product Description
Millipore
SOC Broth, suitable for microbiology
Sigma-Aldrich
Hexammine cobalt(III) chloride, for use in transformations, X-ray crystallography and NMR
Sigma-Aldrich
Hanahan′s Broth (SOB Medium), powder microbial growth medium