MilliporeSigma
  • Characterization of the major laccase isoenzyme from Trametes pubescens and regulation of its synthesis by metal ions.
Characterization of the major laccase isoenzyme from Trametes pubescens and regulation of its synthesis by metal ions.
Microbiology (Reading, England) (2002-07-09)
Christiane Galhaup, Sabine Goller, Clemens K Peterbauer, Josef Strauss, Dietmar Haltrich
ABSTRACT
The major laccase isoenzyme LAP2 secreted by the white-rot basidiomycete Trametes pubescens in response to high copper concentrations was purified to apparent electrophoretic homogeneity using anion-exchange chromatography and gel filtration. The monomeric protein has a molecular mass of 65 kDa, of which 18% is glycosylation, and a pI value of 2.6. The pH optima of the laccase depend on the substrates oxidized and show bell-shaped pH activity profiles with an optimum of 3-4.5 for phenolic substrates such as 2,6-dimethoxyphenol or syringaldazine, while the non-phenolic substrates ABTS [2,2'-azino-bis(3-ethylbenzthiazoline-6-sulfonic acid)] and ferrocyanide show a monotonic pH profile with a rate increasing with decreasing pH. The catalytic efficiencies k(cat)/K(m) determined for some of its substrates were 48 x 10(6), 47 x 10(6), 20 x 10(6) and 7 x 10(6) M(-1) s(-1) for ABTS, syringaldazine, ferrocyanide and oxygen, respectively. Furthermore, the gene lap2 encoding the purified laccase was cloned and its nucleotide sequence determined. The gene consists of 1997 bp, with the coding sequence interrupted by eight introns and flanked by an upstream region in which putative CAAT, TATA, MRE and CreA consensus sequences were identified. Based on Northern analysis containing total RNA from both induced and uninduced cultures, expression of lap2 is highly induced by copper, which is also corroborated by an increase in laccase activity in response to copper. A stimulating effect of various other heavy metal ions on laccase synthesis was also observed. In addition to induction, a second regulatory mechanism seems to be repression of lap2 transcription by glucose.
Product Description
Millipore
Peptone from casein, enzymatic digest, Enzymatic digest of casein, for biotechnological purposes
Sigma-Aldrich
Superdex®, Superdex®75 Prep Grade