MilliporeSigma
  • Chiral Brønsted Acid-Catalyzed Metal-Free Asymmetric Direct Reductive Amination Using 1-Hydrosilatrane.
Chiral Brønsted Acid-Catalyzed Metal-Free Asymmetric Direct Reductive Amination Using 1-Hydrosilatrane.
The Journal of organic chemistry (2019-04-12)
Vladislav Skrypai, Sami E Varjosaari, Fawwaz Azam, Thomas M Gilbert, Marc J Adler
ABSTRACT
The asymmetric direct reductive amination of prochiral ketones with aryl amines using 1-hydrosilatrane with a chiral Brønsted acid catalyst is reported. This is the first known example of chiral Brønsted acid-catalyzed asymmetric reductive amination using a silane as the hydride source. The reaction features a highly practical reducing reagent and proceeds efficiently at room temperature without a specialized reaction setup or equipment to exclude air or moisture. This method provides high conversion and enantiomeric excess up to 84% of the desired chiral secondary amines with minimal side products.
Sigma-Aldrich
1-Hydrosilatrane, ≥95%