MilliporeSigma
  • Potent and selective inhibitors of the TASK-1 potassium channel through chemical optimization of a bis-amide scaffold.
Potent and selective inhibitors of the TASK-1 potassium channel through chemical optimization of a bis-amide scaffold.
Bioorganic & medicinal chemistry letters (2014-07-16)
Daniel P Flaherty, Denise S Simpson, Melissa Miller, Brooks E Maki, Beiyan Zou, Jie Shi, Meng Wu, Owen B McManus, Jeffrey Aubé, Min Li, Jennifer E Golden
ABSTRACT
TASK-1 is a two-pore domain potassium channel that is important to modulating cell excitability, most notably in the context of neuronal pathways. In order to leverage TASK-1 for therapeutic benefit, its physiological role needs better characterization; however, designing selective inhibitors that avoid the closely related TASK-3 channel has been challenging. In this study, a series of bis-amide derived compounds were found to demonstrate improved TASK-1 selectivity over TASK-3 compared to reported inhibitors. Optimization of a marginally selective hit led to analog 35 which displays a TASK-1 IC50=16 nM with 62-fold selectivity over TASK-3 in an orthogonal electrophysiology assay.
Sigma-Aldrich
ML365, ≥98% (HPLC)