MilliporeSigma
  • An in vitro inhibitory effect on RAW 264.7 cells by anti-inflammatory compounds from Smilax corbularia Kunth.
An in vitro inhibitory effect on RAW 264.7 cells by anti-inflammatory compounds from Smilax corbularia Kunth.
Asian Pacific journal of allergy and immunology (2013-02-12)
Srisopa Ruangnoo, Nuanjan Jaiaree, Sunita Makchuchit, Sumalee Panthong, Pakakrong Thongdeeying, Arunporn Itharat
ABSTRACT
Smilax corbularia is a Thai medicinal plant locally known as 'Hua-Khao-Yen Neua', which is used for treating inflammatory conditions. To evaluate the anti-inflammatory effect of S. corbularia extracts and its isolated compounds by determination of inhibitory effects on lipopolysaccharide-stimulated PGE2 release, and TNF-alpha and NO production from RAW 264.7 cells. The inhibitory effect of aqueous and ethanolic extracts of this plant were determined on LPS-induced NO production, TNF-alpha and PGE2 release in RAW 264.7 cells, as an in vitro indication of possible anti-inflammatory activity. The compounds from active extract were isolated by bioassay-guided fractionation. Only the ethanolic extract of this plant inhibited TNF-alpha and NO production, with IC50 values of 61.97, and 83.90 microg/ml respectively. Three flavonols, engeletin, astilbin and quercetin were isolated from the ethanolic extract. quercetin possessed the highest inhibitory effect on NO production with IC50 11.2 microg/ml (37.1 microM), whereas engeletin and astilbin had no activity (IC50 >100 microg/ml). All three flavonols possessed potent inhibition of PGE2 release with IC50 values of 14.4, 19.6 and 19.9 microg/ml (33.2, 43.5 and 65.8 microM) respectively. Quercetin also exhibited the highest inhibitory effect on TNF-alpha production (IC50 = 1.25 microg/ml or 4.14 microM), but engeletin and astilbin had no activity. This is the first report of isolated compounds from S. corbularia with potential anti-inflammatory effects, and the results support the use of this plant by Thai traditional doctors for treatment of inflammatory diseases.
Sigma-Aldrich
Engeletin, ≥98% (HPLC)