MilliporeSigma
  • Biophysical characterisation reveals structural disorder in the nucleolar protein, Dribble.
Biophysical characterisation reveals structural disorder in the nucleolar protein, Dribble.
Biochemical and biophysical research communications (2006-03-18)
C-P Benny Yiu, Rebecca L Beavil, H Y Edwin Chan
ABSTRACT
Dribble (DBE) is an essential protein in Drosophila that belongs to the evolutionarily conserved Krr1p protein family. Proteins in this family are localised in the cell nucleolus and are important for the processing of ribosomal RNAs. However, little is known about their structural and biophysical properties. We have expressed and purified full-length DBE protein from Escherichia coli. Consistent with the native role of DBE in RNA processing, recombinant DBE was shown to bind RNA homo-polymers in vitro. By bioinformatics, size-exclusion chromatography, equilibrium sedimentation analysis, controlled proteolysis, and a variety of spectroscopic techniques, we have found that DBE is a monomeric protein in solution containing both alpha- and beta-structures. Moreover, the structure of DBE is expanded and significantly disordered (approximately 45% disordered). Natively disordered proteins are thought to provide a disproportionately large surface area and structural plasticity for nucleic acid binding. We therefore propose that the presence of structural disorder is an important feature of DBE that facilitates the protein to interact with RNAs in the nucleolus.
Sigma-Aldrich
Polycytidylic acid–Agarose, lyophilized powder