MilliporeSigma
  • Isolation and characterization of a novel ribonuclease from the pink oyster mushroom Pleurotus djamor.
Isolation and characterization of a novel ribonuclease from the pink oyster mushroom Pleurotus djamor.
The Journal of general and applied microbiology (2010-07-22)
Xiangli Wu, Suyue Zheng, Li Cui, Hexiang Wang, Tzi Bun Ng
ABSTRACT
A 15-kDa RNase was purified from Pleurotus djamor using ion exchange chromatography and gel filtration. Its N-terminal amino acid sequence was different from previously reported RNase sequences of mushrooms belonging to the genus of Pleurotus and other genera. The RNase exhibited maximal RNase activity at pH 4.6 and 60 degrees C. Its activity toward polyhomoribonucleotides was poly(U) > poly(C) > poly(A) > poly(G). It inhibited proliferation of hepatoma cells and breast cancer cells. The ranking of inhibitory potencies of metal ions on RNase activity was Fe(3+)> Al(3+)> Ca(2+)> Hg(2+). The isolated RNase had a distinctive N-terminal sequence and optimum pH. It exhibited antiproliferative activity on tumor cells.
Sigma-Aldrich
Ribonucleic acid from baker′s yeast (S. cerevisiae)