MilliporeSigma
  • Crystallization and preliminary X-ray analysis of PaaAC, the main component of the hydroxylase of the Escherichia coli phenylacetyl-coenzyme A oxygenase complex.
Crystallization and preliminary X-ray analysis of PaaAC, the main component of the hydroxylase of the Escherichia coli phenylacetyl-coenzyme A oxygenase complex.
Acta crystallographica. Section F, Structural biology and crystallization communications (2010-09-09)
Andrey M Grishin, Eunice Ajamian, Linhua Zhang, Miroslaw Cygler
ABSTRACT
The Escherichia coli paa operon encodes enzymes of the phenylacetic acid-utilization pathway that metabolizes phenylacetate in the form of a coenzyme A (CoA) derivative. The phenylacetyl-coenzyme A oxygenase complex, which has been postulated to contain five components designated PaaABCDE, catalyzes ring hydroxylation of phenylacetyl-CoA. The PaaAC subcomplex shows low sequence similarity to other bacterial multicomponent monooxygenases (BMMs) and forms a separate branch on the phylogenetic tree. PaaAC, which catalyzes the hydroxylation reaction, was purified and crystallized in the absence of a bound ligand as well as in complexes with CoA, 3-hydroxybutyryl-CoA, benzoyl-CoA and the true substrate phenylacetyl-CoA. Crystals of the ligand-free enzyme belonged to space group P2(1)2(1)2(1) and diffracted to 2.65 A resolution, whereas complexes with CoA and its derivatives crystallized in space group P4(1)2(1)2 and diffracted to approximately 2.0 A resolution. PaaAC represents the first crystallized BMM hydroxylase that utilizes a CoA-linked substrate.
Sigma-Aldrich
Phenylacetyl coenzyme A lithium salt, ~95%