- Ligand-Enabled β-C(sp3)-H Olefination of Free Carboxylic Acids.
Ligand-Enabled β-C(sp3)-H Olefination of Free Carboxylic Acids.
Journal of the American Chemical Society (2018-07-22)
Zhe Zhuang, Chang-Bin Yu, Gang Chen, Qing-Feng Wu, Yi Hsiao, Candice L Joe, Jennifer X Qiao, Michael A Poss, Jin-Quan Yu
PMID30029574
ABSTRACT
An acetyl-protected aminoethyl phenyl thioether has been developed to promote C(sp3)-H activation. Significant ligand enhancement is demonstrated by the realization of the first Pd(II)-catalyzed olefination of C(sp3)-H bonds of free carboxylic acids without using an auxiliary. Subsequent lactonization of the olefinated product via 1,4 addition provided exclusively monoselectivity in the presence of multiple β-C-H bonds. The product γ-lactone can be readily opened to give either the highly valuable β-olefinated or γ-hydroxylated aliphatic acids. Considering the challenges in developing Heck couplings using alkyl halides, this reaction offers a useful alternative.
MATERIALS