MilliporeSigma
  • Realizing Small Energy Loss of 0.55 eV, High Open-Circuit Voltage >1 V and High Efficiency >10% in Fullerene-Free Polymer Solar Cells via Energy Driver.
Realizing Small Energy Loss of 0.55 eV, High Open-Circuit Voltage >1 V and High Efficiency >10% in Fullerene-Free Polymer Solar Cells via Energy Driver.
Advanced materials (Deerfield Beach, Fla.) (2017-01-20)
Pei Cheng, Mingyu Zhang, Tsz-Ki Lau, Yao Wu, Boyu Jia, Jiayu Wang, Cenqi Yan, Meng Qin, Xinhui Lu, Xiaowei Zhan
ABSTRACT
A new, easy, and efficient approach is reported to enhance the driving force for charge transfer, break tradeoff between open-circuit voltage and short-circuit current, and simultaneously achieve very small energy loss (0.55 eV), very high open-circuit voltage (>1 V), and very high efficiency (>10%) in fullerene-free organic solar cells via an energy driver.
Sigma-Aldrich
IDT-2Br, ≥99%