MilliporeSigma
  • Nicotinamide adenine dinucleotide species in the horseradish peroxidase-oxidase oscillator.
Nicotinamide adenine dinucleotide species in the horseradish peroxidase-oxidase oscillator.
European journal of biochemistry (2000-08-10)
E S Kirkor, A Scheeline
ABSTRACT
NADH chemistry ancillary to the oscillatory peroxidase-oxidase (PO) reaction has been reexamined. Previously, (NAD)2 has been thought of as a terminal, inert product of the PO reaction. We now show that (NAD)2 is a central reactant in this system. Although we found traces of the dimer after several hours of the PO reaction, no accumulation of the dimer occurred, regardless of the reaction time or the number of oscillations. (NAD)2 can convert horseradish peroxidase (HRP) compound I (CpI) to compound II (CpII) with apparent rate constant (2.7 +/- 0.2) x 105 M-1.s-1 and CpII to HRP at 1 x 105 M-1.s-1. Moreover, a reduction of HRP compound III (CpIII) to CpI by (NAD)2 occurs with a rate constant faster than 5 x 106 M-1.s-1. The (NAD)2 reduction of CpIII provides an alternative to the reduction by NAD radical suggested by Yokota and Yamazaki. HRP catalyzes oxidation of alpha-NADH, not only the beta anomer as previously assumed. Rate constants of alpha- and beta-NADH reactions with CpI are (7.4 +/- 0.4) x 105 M-1.s-1, and (1.7 +/- 0.2) x 105 M-1.s-1, and with CpII are estimated as 5 x 104 M-1.s-1, and 4 x 104 M-1.s-1. Apparent rate constants of reduction of methylene blue (MB) to leuco-methylene blue (MBH) are 3.8 x 104 M-1.s-1 for NADH and 6.4 x 104 M-1.s-1 for NAD dimer, (NAD)2, while reoxidation of MBH proceeds at (2.1 +/- 0.2) x 103 M-1.s-1 All the rates were measured in 0.1 M acetate buffer, pH 5.1.
Sigma-Aldrich
α-Nicotinamide adenine dinucleotide, reduced disodium salt, ≥90%