MilliporeSigma
  • Conformational variability of poly(dA-dT).poly(dA-dT) and some other deoxyribonucleic acids includes a novel type of double helix.
Conformational variability of poly(dA-dT).poly(dA-dT) and some other deoxyribonucleic acids includes a novel type of double helix.
Journal of biomolecular structure & dynamics (1985-08-01)
M Vorlícková, J Kypr
ABSTRACT
The article reviews data indicating that poly(dA-dT).poly(dA-dT) is able of adopting three distinct double helical structures in solution, of which only the A form conforms to classical notions. The other two structures have dinucleotides as double helical repeats. At low salt concentrations poly(dA-dT).poly(dA-dT) adopts a B-type alternating conformation which is exceptionally variable. Its architecture can gradually move in the limits demarcated by the CD spectra with inverted long wavelength CD bands and the 31P NMR spectra with a very low and a 0.6 ppm separation of two resonances. Contrary to Z-DNA, the 31P NMR spectrum of the limiting alternating B conformation of poly(dA-dT).poly(dA-dT) is characterized by an upfield shift of one resonance. We attribute the exceptional conformational flexibility of the alternating B conformation to the unequal tendency of bases in the dA-dT and dT-dA steps to stack. However, by assuming the limiting alternating B conformation, the variability of the synthetic DNA is not exhausted. Specific agents make it isomerize into another conformation by a fast, two-state mechanism, which is reflected by a further deepening of the negative long wavelength CD band and a downfield shift of the 31P NMR resonance of poly(dA-dT).poly(dA-dT) that was constant in the course of the gradual alterations of the alternating B conformation. These changes are, however, qualitatively different from the way poly(dG-dC).poly(dG-dC) behaves in the course of the B-Z isomerization. Poly(dG-dC).poly(dG-dC) displays purine-pyrimidine (dGpdC) resonance in the characteristic downfield position, while the downfield resonance of poly(dA-dT).poly(dA-dT) belongs to the pyrimidine-purine (dTpdA) phosphodiester linkages. Consequently, phosphodiester linkages in the purine-pyrimidine steps play a similar role in the appearance of the Z form to the pyrimidine-purine phosphodiesters in the course of the isomerization of poly(dA-dT).poly(dA-dT). This excludes that the high-salt structures of poly(dA-dT).poly(dA-dT) and poly(dG-dC).poly(dG-dC) are members of the same conformational family. We call the high-salt conformation of poly(dA-dT).poly(dA-dT) X-DNA. It furthermore follows from the review that synthetic molecules of DNA with alternating purine-pyrimidine sequences of bases can adopt either the Z form or the X form, or even both, depending on the environmental conditions. This introduces a new dimension into the DNA double helix conformational variability. The possible biological relevance of the X form is suggested by experiments with linear molecules of natural DNA.(ABSTRACT TRUNCATED AT 400 WORDS)
Sigma-Aldrich
Polydeoxyadenylic acid • Polythymidylic acid sodium salt, double-stranded homopolymer
Sigma-Aldrich
Poly(deoxyadenylic-thymidylic) acid sodium salt, double-stranded alternating copolymer