MilliporeSigma
  • Structure elucidation and antioxidant activity of a novel polysaccharide isolated from Boletus speciosus Forst.
Structure elucidation and antioxidant activity of a novel polysaccharide isolated from Boletus speciosus Forst.
International journal of biological macromolecules (2012-01-31)
Xiang Ding, Yi-ling Hou, Wan-ru Hou
ABSTRACT
In this study, a novel heteropolysaccharide was isolated from the fruiting bodies of Boletus speciosus Forst through DEAE-cellulose column and Sephadex G-200 column. The Boletus speciosus Forst polysaccharide (BSFP-1) had a molecular weight of 1.33×10(4) Da and was mainly composed of l-Man and d-Gal which ratios were 2:1. Structural features of Boletus speciosus Forst polysaccharide (BSFP-1) were investigated by a combination of total hydrolysis, methylation analysis, gas chromatography-mass spectrometry (GC-MS), infrared (IR) spectra and nuclear magnetic resonance (NMR) spectroscopy. The results indicated that Boletus speciosus Forst polysaccharide (BSFP-1) had a backbone of (1→4)-α-l-mannopyranose residues which branches at O-6 based on the experimental results. The branches were mainly composed of one with →1)-α-d-galactopyranose residue. The antioxidant activity of BSFP-1 was evaluated with two biochemical methods, including 1,1-diphenyl-2-picrylhydrazyl (DPPH(-)) radical scavenging, scavenging activity of 2,2'-azino-bis(3-ethylbenzthiazoline-6-suphonic acid)diammonium (ABTS(+)) radical. The results indicated that BSFP-1 showed strong antioxidant.