MilliporeSigma
  • Bio-orthogonal labeling as a tool to visualize and identify newly synthesized proteins in Caenorhabditis elegans.
Bio-orthogonal labeling as a tool to visualize and identify newly synthesized proteins in Caenorhabditis elegans.
Nature protocols (2014-08-29)
Milena Ullrich, Vanessa Liang, Yee Lian Chew, Samuel Banister, Xiaomin Song, Thiri Zaw, Hong Lam, Slavica Berber, Michael Kassiou, Hannah R Nicholas, Jürgen Götz
ABSTRACT
In this protocol we describe the incorporation of bio-orthogonal amino acids as a versatile method for visualizing and identifying de novo-synthesized proteins in the roundworm Caenorhabditis elegans. This protocol contains directions on implementing three complementary types of analysis: 'click chemistry' followed by western blotting, click chemistry followed by immunofluorescence, and isobaric tags for relative and absolute quantification (iTRAQ) quantitative mass spectrometry. The detailed instructions provided herein enable researchers to investigate the de novo proteome, an analysis that is complicated by the fact that protein molecules are chemically identical to each other, regardless of the timing of their synthesis. Our protocol circumvents this limitation by identifying de novo-synthesized proteins via the incorporation of the chemically modifiable azidohomoalanine instead of the natural amino acid methionine in the nascent protein, followed by facilitating the visualization of the resulting labeled proteins in situ. It will therefore be an ideal tool for studying de novo protein synthesis in physiological and pathological processes including learning and memory. The protocol requires 10 d for worm growth, liquid culture and synchronization; 1-2 d for bio-orthogonal labeling; and, with regard to analysis, 3-4 d for western blotting, 5-6 d for immunofluorescence or ~3 weeks for mass spectrometry.
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, for molecular biology, 99.0-101.0% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Goat serum
Sigma-Aldrich
DAPI, for nucleic acid staining
Millipore
Benzonase® Nuclease, ≥250 units/μL, ≥90% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution