MilliporeSigma
  • Identification of a dominant negative functional domain on DAPK-1 that degrades DAPK-1 protein and stimulates TNFR-1-mediated apoptosis.
Identification of a dominant negative functional domain on DAPK-1 that degrades DAPK-1 protein and stimulates TNFR-1-mediated apoptosis.
The Journal of biological chemistry (2007-02-28)
Yao Lin, Craig Stevens, Ted Hupp
ABSTRACT
DAPK-1 is a stress-activated tumor suppressor protein that plays a role in both proapoptotic or antiapoptotic signal transduction pathways. To define mechanisms of DAPK-1 protein regulation, we have determined that DAPK-1 protein has a long half-life, and therefore its activity is primarily regulated at the protein level. Changes in DAPK-1 protein levels occur by a cathepsin B-dependent pathway, prompting us to evaluate whether cathepsin B plays positive or negative role in DAPK-1 function. The transfection of p55-TNFR-1 induced complex formation between DAPK-1 and cathepsin B. Depletion of cathepsin B protein using small interfering RNA stimulated TNFR-1 dependent apoptosis. The minimal binding region on DAPK-1 for cathepsin B was mapped to amino acids 836-947. The transfection of the DAPK-1-(836-947) miniprotein acted in a dominant negative manner inducing endogenous DAPK-1 protein degradation in a TNFR-1-dependent manner. These data suggest that DAPK-1 forms a multiprotein survival complex with cathepsin B countering the rate of TNFR-1-dependent apoptosis and highlights the importance of developing DAPK-1 inhibitors as agents to sensitize cells to stress-induced apoptosis.
Sigma-Aldrich
ApopTag Plus In Situ Apoptosis Fluorescein Detection Kit, The ApopTag Plus Fluorescein In Situ Apoptosis Detection Kit detects apoptotic cells in situ by the indirect TUNEL method, utilizing an anti-digoxigenin antibody that is conjugated to a fluorescein reporter molecule.