MilliporeSigma
  • Purification, cloning, and functional expression of dihydroneopterin triphosphate 2'-epimerase from Escherichia coli.
Purification, cloning, and functional expression of dihydroneopterin triphosphate 2'-epimerase from Escherichia coli.
The Journal of biological chemistry (1997-06-13)
C Ahn, J Byun, J Yim
ABSTRACT
Dihydroneopterin triphosphate (H2NTP) 2'-epimerase from Escherichia coli catalyzes the epimerization of H2NTP to dihydromonapterin triphosphate (H2MTP). The enzyme was purified 954-fold to apparent homogeneity by a combination of ammonium sulfate fractionation and column chromatography of Cibacron blue 3GA dye ligand, phenyl-Sepharose CL-4B, methotrexate-agarose, and Superdex 200 HR 10/30 FPLC column. The molecular mass of the epimerase determined on a Superdex column was 82.6 kDa, while the subunit molecular mass determined on SDS-polyacrylamide gel electrophoresis was 13.7 kDa. This implies that the epimerase most probably exists as homohexamer. The 20-amino acid sequence from the N terminus was determined (AQPAAIIRIKNLRLRTFIGI). Based on this sequence, the gene encoding the epimerase was cloned using a simple polymerase chain reaction approach. Translation of the nucleotide sequence of the cloned gene revealed the presence of an open reading frame containing 120 amino acids with a predicted molecular mass of 13,993 Da. The epimerase gene located in a 2.3-kilobase BamHI-EcoRI fragment from Kohara's clone 406 was overexpressed 300-fold, which was confirmed by the prominent increase in the 14-kDa protein band on SDS-polyacrylamide electrophoresis gels. It showed no homology with the sequences of isomerases or other enzymes in GenBank/EMBL data bases.
Sigma-Aldrich
Methotrexate−Agarose, saline suspension