MilliporeSigma
Pitavastatin: A Review in Hypercholesterolemia.
American journal of cardiovascular drugs : drugs, devices, and other interventions (2017-01-29)
Sheridan M Hoy
ABSTRACT
Oral pitavastatin (Livalo®; Livazo®) is a competitive HMG-CoA reductase inhibitor that is available in the EU for the reduction of elevated total cholesterol and low-density lipoprotein cholesterol (LDL-C) levels in adults with primary hypercholesterolemia and combined (mixed) dyslipidemia. In short-term, phase III or IV studies in this patient population, pitavastatin 1-4 mg once daily was generally no less effective than presumed equipotent dosages of atorvastatin and simvastatin (including in patients with type 2 diabetes or ≥2 cardiovascular risk factors) and was superior to pravastatin (including in patients aged ≥65 years) in lowering LDL-C levels. Pitavastatin provided sustained LDL-C-lowering efficacy over up to 60 weeks' therapy in extension studies, and was associated with short- and longer-term improvements in several other lipid parameters. Short- and longer-term outcomes in studies in Asian patients were consistent with these findings. Pitavastatin was generally well tolerated and did not appear to adversely affect glucose metabolism parameters (e.g. fasting blood glucose, fasting plasma glucose, fasting plasma insulin, glycated hemoglobin) in short- and longer-term prospective and post-marketing surveillance studies in adults. Moreover, in combination with lifestyle modification advice, it was associated with a significant reduction in the risk of progression from impaired glucose tolerance to diabetes relative to lifestyle modification advice alone in a longer-term study in Japanese subjects. Thus, pitavastatin is an effective treatment option in adults with primary hypercholesterolemia and combined (mixed) dyslipidemia, including those at risk of developing type 2 diabetes.
Sigma-Aldrich
Pitavastatin calcium, ≥98% (HPLC)