MilliporeSigma
  • Neurabin-II/spinophilin. An actin filament-binding protein with one pdz domain localized at cadherin-based cell-cell adhesion sites.
Neurabin-II/spinophilin. An actin filament-binding protein with one pdz domain localized at cadherin-based cell-cell adhesion sites.
The Journal of biological chemistry (1998-03-07)
A Satoh, H Nakanishi, H Obaishi, M Wada, K Takahashi, K Satoh, K Hirao, H Nishioka, Y Hata, A Mizoguchi, Y Takai
ABSTRACT
In a preceding paper, we reported a novel actin filament (F-actin)-binding protein, named neurabin, which was specifically expressed in neural tissue and implicated in neurite formation. We purified from rat brain another F-actin-binding protein, which had a domain organization similar to that of neurabin but was ubiquitously expressed, and named it neurabin-II. The original neurabin, renamed neurabin-I, had 1095 amino acids and a calculated Mr of 122,729, whereas neurabin-II had 817 amino acids and a calculated Mr of 89, 642. Both neurabin-I and -II had one F-actin-binding domain at the N-terminal region, one PDZ domain at the middle region, a domain known to interact with transmembrane proteins, and domains predicted to form coiled-coil structures at the C-terminal region. Both neurabin-I and -II bound along the sides of F-actin and showed F-actin-cross-linking activity. The subcellular distribution analysis indicated that neurabin-II was enriched at the postsynaptic density fraction in rat brain and the adherens junction fraction in rat liver. Immunofluorescence microscopic analysis revealed that neurabin-II was highly concentrated at the synapse in primary cultured rat hippocampal neurons and at the cadherin-based cell-cell adhesion sites in Madin-Darby canine kidney cells. Neurabin-II turned out to be the same as a recently reported protein phosphatase 1-binding protein named spinophilin. These results suggest that neurabin-II/spinophilin plays an important role in linking the actin cytoskeleton to the plasma membrane.