MilliporeSigma
  • The Xenopus Wnt effector XTcf-3 interacts with Groucho-related transcriptional repressors.
The Xenopus Wnt effector XTcf-3 interacts with Groucho-related transcriptional repressors.
Nature (1998-10-23)
J Roose, M Molenaar, J Peterson, J Hurenkamp, H Brantjes, P Moerer, M van de Wetering, O Destrée, H Clevers
ABSTRACT
Tcf/Lef transcription factors mediate signalling from Wingless/Wnt proteins by recruiting Armadillo/beta-catenin as a transcriptional co-activator. However, studies of Drosophila, Xenopus and Caenorhabditis elegans have indicated that Tcf factors may also be transcriptional repressors. Here we show that Tcf factors physically interact with members of the Groucho family of transcriptional repressors. In transient transfection assays, the Xenopus Groucho homologue XGrg-4 inhibited activation of transcription of synthetic Tcf reporter genes. In contrast, the naturally truncated Groucho-family member XGrg-5 enhanced transcriptional activation. Injection of XGrg-4 into Xenopus embryos repressed transcription of Siamois and Xnr-3, endogenous targets of beta-catenin-Tcf. Dorsal injection of XGrg-4 had a ventralizing effect on Xenopus embryos. Secondary-axis formation induced by a dominant-positive Armadillo-Tcf fusion protein was inhibited by XGrg-4 and enhanced by XGrg-5. These data indicate that expression of Tcf target genes is regulated by a balance between Armadillo and Groucho.
Sigma-Aldrich
TCF Reporter Plasmid Kit, Set of transfection grade T cell factor (TCF) reporter plasmids for use in TOPFlash and FOPFlash wnt/b-catenin activity assays.
Sigma-Aldrich
FOPflash (mutant TCF binding sites), Transfection grade T cell factor (TCF) reporter plasmid containing 2 full & one incomplete copy of the Tcf binding site (mutated) followed by 3 copies in the reverse orientation. Serves as a negative control to TOPflash.
Sigma-Aldrich
TOPflash (TCF Reporter Plasmid), Transfection grade T-cell factor reporter plasmid containing 2 sets (with the second set in the reverse orientation) of 3 copies of the TCF binding site (wild type) upstream of the Thymidine Kinase minimal promoter and Luciferase open reading frame.