MilliporeSigma
  • Selective expression of a novel surface molecule by human Th2 cells in vivo.
Selective expression of a novel surface molecule by human Th2 cells in vivo.
Journal of immunology (Baltimore, Md. : 1950) (1999-02-11)
K Nagata, K Tanaka, K Ogawa, K Kemmotsu, T Imai, O Yoshie, H Abe, K Tada, M Nakamura, K Sugamura, S Takano
ABSTRACT
The search for reliable marker molecules discriminating between human Th1 and Th2 cells identified a gene encoding a novel member of the G protein-coupled leukocyte chemoattractant receptor family, which is selectively expressed in Th2 but not Th1 lineage cells, thereby named CRTH2 (chemoattractant receptor-homologous molecule expressed on Th2 cells). Studies with anti-CRTH2 mAbs demonstrated that CRTH2 was expressed in a small population (0.4-6.5%) of CD4+ T cells in fresh PBMCs of healthy adults, but no remarkable expression was seen in B cells and NK cells. In some cases, CD8+ T cells ( approximately 3.5%) expressed CRTH2. Phenotypes of CD4+ T cells expressing CRTH2 were CD45RA-, CD45RO+, and CD25+, similar to those of Ag-activated effector/memory T cells. Freshly isolated CRTH2+ CD4+ T cells produced Th2- but little or no Th1-type cytokines upon stimulation with PMA and ionomycin. In addition, an allergen-induced proliferative response in fresh PBMCs was significantly and selectively reduced by subtracting CRTH2+ cells. Together, these results indicate that CRTH2 is selectively expressed in vivo in an activated state of Th2 cells including allergen-responsive Th2 cells, suggesting its pivotal roles in ongoing Th2-type immune reactions.
Sigma-Aldrich
ChemiSCREEN CRTH2 Membrane Preparation, Human CRTH2 / DP2 / GPR44 GPCR membrane preparation for Radioligand binding Assays & GTPgammaS binding.