MilliporeSigma
  • Somatic mutations of the epidermal growth factor receptor and non-small-cell lung cancer.
Somatic mutations of the epidermal growth factor receptor and non-small-cell lung cancer.
Journal of medical genetics (2006-12-13)
Xiaozhu Zhang, Alex Chang
ABSTRACT
Frequent overexpression of epidermal growth factor receptor (EGFR) in non-small-cell lung cancer (NSCLC) makes EGFR a new therapeutic target. Two specific EGFR tyrosine kinase inhibitors, gefitinib (ZD1839, Iressa) and erlotinib (OSI-774, Tarceva), have been developed and approved by the US Food and Drug Administration for second-line and third-line treatment of advanced NSCLC. Clinical trials have shown considerable variability in the response rate between different patients with NSCLC, which led to the discovery of somatic EGFR-activating mutations. This brief review summarises the discovery and functional consequences of the mutations, their clinicopathological features and significant implications in the treatment and prognosis of NSCLC.