MilliporeSigma
Tip60 acetyltransferase activity is controlled by phosphorylation.
The Journal of biological chemistry (2002-12-07)
Claudie Lemercier, Gaëlle Legube, Cécile Caron, Mathilde Louwagie, Jérôme Garin, Didier Trouche, Saadi Khochbin
ABSTRACT
Here we show that the phosphorylation of histone acetyltransferase Tip60, a target of human immunodeficiency virus, type 1-encoded transactivator Tat, plays a crucial role in the control of its catalytic activity. Baculovirus-based expression and purification of Tip60 combined with mass spectrometry allowed the identification of serines 86 and 90 as two major sites of phosphorylation in vivo. The phosphorylation of Tip60 was found to modulate its histone acetyltransferase activity. One of the identified phosphorylated serines, Ser-90, was within a consensus cyclin B/Cdc2 site. Ser-90 was specifically phosphorylated in vitro by the cyclin B/Cdc2 complex. Accordingly, the phosphorylation of Tip60 was enhanced after drug-induced arrest of cells in G(2)/M. This G(2)/M-dependent phosphorylation of Tip60 was abolished by treating cells with a specific inhibitor of the cyclin-dependent kinase, roscovitin. All together, these results strongly suggest a G(2)/M-dependent control of Tip60 activity.
Sigma-Aldrich
Phosphatase, Alkaline−Agarose from calf intestine, ammonium sulfate suspension