MilliporeSigma
Resonance Raman investigation of nickel microperoxidase-11.
Biochemistry (1999-03-03)
J G Ma, J M Vanderkooi, J Zhang, S L Jia, J A Shelnutt
ABSTRACT
Resonance Raman and UV-visible absorption spectra show that nickel(II) microperoxidase-11 (NiMP-11) is four-coordinate in aqueous solution in the pH range from 1.0 to 13.0. In aqueous solutions of NiMP-11 in the absence of cetyltrimethylammonium bromide (CTAB), NiMP-11 is aggregated. In CTAB micellar solutions, where aggregation of NiMP-11 does not occur, the Raman spectra of NiMP-11 are similar to that of nickel(II) cytochrome c (NiCyt-c). The presence of the peptide segment shifts the equilibrium heavily in favor of the nonplanar form, just as does the entire protein component in the case of NiCyt-c. This further elucidates the structural mechanism by which the protein segment ruffles the heme, most likely modulating the redox potential as indicated for the cytochromes c3 [Ma, J.-G., et al. (1998) Biochemistry 37, 12431-12442]. Furthermore, the hydrophobic environment that is provided by the CTAB micelle is found to be crucial to the native folding of the pentapeptide and formation of two hydrogen bonds in the peptide backbone. These two H-bonds act to contract the peptide segment exerting the force on the macrocycle that causes the ruffling and makes the redox potential more negative than if the heme were to remain planar. The structure of the heme and pentapeptide may also be associated with redox-linked triggering of the formation and release of cytochrome-protein complexes.
Sigma-Aldrich
Microperoxidase (MP-11) sodium salt, ≥85% (HPLC)