MilliporeSigma
  • The serine/threonine kinase Pim-2 is a transcriptionally regulated apoptotic inhibitor.
The serine/threonine kinase Pim-2 is a transcriptionally regulated apoptotic inhibitor.
Genes & development (2003-07-19)
Casey J Fox, Peter S Hammerman, Ryan M Cinalli, Stephen R Master, Lewis A Chodosh, Craig B Thompson
ABSTRACT
Growth factor withdrawal results in the termination of factor-dependent transcription. One transcript that declines rapidly following growth factor deprivation of hematopoietic cells is the serine/threonine kinase pim-2. When constitutively expressed, Pim-2 conferred long-term resistance to a variety of apoptotic stimuli including growth factor withdrawal and endogenous levels of Pim-2 contributed to growth factor-mediated apoptotic resistance. Pim-2 expression maintained cell size and mitochondrial potential independently of the PI3K/Akt/TOR pathway. Pim-2-dependent maintenance of cell size and survival correlated with its ability to maintain rapamycin-resistant phosphorylation of the translational repressor 4E-BP1 and phosphorylation of the BH3 protein BAD. These results establish Pim-2 as a direct link between growth factor-induced transcription and a novel, kinase-dependent pathway that promotes cell-autonomous survival.
Sigma-Aldrich
PIM2 Protein, active, 25 µg, Active, human PIM2 expressed in E. coli, purified using GSH agarose, for use in Kinase Assays.