MilliporeSigma
CrkL directs ASAP1 to peripheral focal adhesions.
The Journal of biological chemistry (2003-01-11)
Atsushi Oda, Ikuo Wada, Koichi Miura, Katsuya Okawa, Toshihiko Kadoya, Takashi Kato, Hiroshi Nishihara, Masae Maeda, Shinya Tanaka, Kazuo Nagashima, Chiaki Nishitani, Kazuhiko Matsuno, Masaho Ishino, Laura M Machesky, Hiroyoshi Fujita, Paul Randazzo
ABSTRACT
Searching for proteins in platelets that can interact with the N-terminal SH3 domain of CrkL (using a combination of a pull-down assay followed by mass spectrometry), we have found that human platelets express an ADP-ribosylation factor (Arf)-specific GTPase-activating protein (GAP), ASAP1, as a CrkL-binding protein. In spreading platelets, most endogenous ASAP1 is localized at peripheral focal adhesions. To determine the physiologic significance of the CrkL-ASAP1 association, we overexpressed CrkL, ASAP1, or both in combination in COS7 cells. Unlike endogenous ASAP1 in platelets, overexpressed ASAP1 showed diffuse cytoplasmic distribution. However, when co-expressed with wild-type CrkL, both endogenous and expressed ASAP1 accumulated at CrkL-induced focal adhesions. An SH2-mutated CrkL, which cannot localize at focal adhesions, failed to recruit ASAP1 into focal adhesions. Thus, CrkL appears to be a lynchpin between ASAP1 and peripheral focal adhesions.