MilliporeSigma
Role of Dok1 in cell signaling mediated by RET tyrosine kinase.
The Journal of biological chemistry (2002-06-28)
Hideki Murakami, Yumiko Yamamura, Yohei Shimono, Kumi Kawai, Kei Kurokawa, Masahide Takahashi
ABSTRACT
Using a yeast two-hybrid screen, we identified Dok1 as a docking protein for RET tyrosine kinase. Dok1 bound more strongly to RET with a multiple endocrine neoplasia (MEN) 2B mutation than RET with a MEN2A mutation and was highly phosphorylated in the cells expressing the former mutant protein. Analysis by site-directed mutagenesis revealed that tyrosine 361 in mouse Dok1 represents a binding site for the Nck adaptor protein and tyrosines 295, 314, 361, 376, 397, and 408 for the Ras-GTPase-activating protein. We replaced tyrosine 361 or these six tyrosines with phenylalanine (designated Y361F or 6F) in Dok1 and introduced the mutant Dok1 genes into the cells expressing the wild-type RET or RET-MEN2B protein. Overexpression of Dok1 or Dok1-Y361F, but not Dok1-6F, suppressed the Ras/Erk activation induced by glial cell line-derived neurotrophic factor or RET-MEN2B, implying that this inhibitory effect requires the Ras-GTPase-activating protein binding to Dok1. In contrast, overexpression of Dok1, but not Dok1-Y361F or Dok1-6F, enhanced the c-Jun amino-terminal kinase (JNK) and c-Jun activation. This suggested that the association of Nck to tyrosine 361 in Dok1 is necessary for the JNK and c-Jun activation by glial cell line-derived neurotrophic factor or RET-MEN2B. Because a high level of the JNK phosphorylation was observed in the cells expressing RET-MEN2B, its strong activation via Nck binding to Dok1 may be responsible for aggressive properties of medullary thyroid carcinoma developed in MEN 2B.