MilliporeSigma
Effect of adding exercise to a diet containing glucomannan.
Metabolism: clinical and experimental (2007-07-10)
William J Kraemer, Jakob L Vingren, Ricardo Silvestre, Barry A Spiering, Disa L Hatfield, Jen Y Ho, Maren S Fragala, Carl M Maresh, Jeff S Volek
ABSTRACT
The aim of this study was to examine the effect of adding a total-body exercise program to an 8-week diet supplemented with glucomannan on weight loss, body composition, blood parameters, and physical performance in overweight men and women. Sedentary men and women who were overweight (body mass index>25 kg m(-2)) (men: 18-57 years, 27.0+/-11.2 years, 177.5+/-7.6 cm, 102.4+/-14.9 kg; women: 18-52 years, 33.4+/-12.1 years, 160.6+/-4.6 cm, 79.9+/-9.3 kg) completed an 8-week diet with 3000 mg glucomannan combined with either no exercise (No-Ex) (10 men, 10 women) or a resistance and endurance exercise training program (Ex) (12 men, 10 women). The diet emphasized healthy food choices and portion size control. The exercise training consisted of 3 weekly sessions of approximately 1 hour of a nonlinear periodized total-body resistance exercise program followed by 30 minutes of endurance exercise. After the intervention, there were reductions (P<.05) in body mass (men, -2.7+/-1.4 and -3.0+/-4.0 kg; women, -2.2+/-1.5 and -3.3+/-1.5 kg; No-Ex and Ex, respectively), fat mass (men, -2.3+/-1.6 and -3.9+/-2.5 kg; women, -2.6+/-1.4 and -3.6+/-1.1 kg; No-Ex and Ex, respectively), total cholesterol (TC) (men, -17.9+/-21.5 and -18.8+/-19.4 mg dL(-1); women, -9.3+/-20.0 and -10.1+/-19.5 mg dL(-1); No-Ex and Ex, respectively), and low-density lipoprotein cholesterol. Exercise significantly improved high-density lipoprotein cholesterol (HDL-C) (No-Ex, -2.0+/-4.7 and -2.3+/-4.5 mg dL(-1) vs Ex, 4.4+/-10.8 and 1.6+/-3.6 mg dL(-1); men and women, respectively), TC/HDL-C ratio, squat and bench press 1-repetition maximum, and distance covered during a shuttle-run test. In addition, exercise appeared to augment the reduction in fat mass (by 63% and 50%; men and women, respectively) and waist circumference, but did not affect total weight loss. Addition of a resistance and endurance exercise training program to a glucomannan diet regimen significantly improved measures of body composition, HDL-C, and TC/HDL-C ratio.
Product Description
Millipore
Human Insulin ELISA, This Human Insulin ELISA is used to measure & quantify insulin levels in Metabolism & Endocrine research.
Pricing and availability is not currently available.