MilliporeSigma
  • Mesoporous silica-supported lipid bilayers (protocells) for DNA cargo delivery to the spinal cord.
Mesoporous silica-supported lipid bilayers (protocells) for DNA cargo delivery to the spinal cord.
Journal of controlled release : official journal of the Controlled Release Society (2013-03-23)
Ellen C Dengler, Juewen Liu, Audra Kerwin, Sergio Torres, Clara M Olcott, Brandi N Bowman, Leisha Armijo, Katherine Gentry, Jenny Wilkerson, James Wallace, Xingmao Jiang, Eric C Carnes, C Jeffrey Brinker, Erin D Milligan
ABSTRACT
Amorphous mesoporous silica nanoparticles ('protocells') that support surface lipid bilayers recently characterized in vitro as carrier constructs for small drug and DNA delivery are reported here as highly biocompatible both in vitro and in vivo, involving the brain and spinal cord following spinal delivery into the lumbosacral subarachnoid space (intrathecal; i.t.). Specifically, positively charged, 1, 2-Dioleoyl-3-Trimethylammonium-Propane (DOTAP)-cholesterol (DOTAP:Chol) liposome-formulated protocells revealed stable in vitro cargo release kinetics and cellular interleukin-10 (IL-10) transgene transfection. Recent approaches using synthetic non-viral vector platforms to deliver the pain-suppressive therapeutic transgene, IL-10, to the spinal subarachnoid space have yielded promising results in animal models of peripheral neuropathy, a condition involving aberrant neuronal communication within sensory pathways in the nervous system. Non-viral drug and gene delivery protocell platforms offer potential flexibility because cargo release-rates can be pH-dependent. We report here that i.t. delivery of protocells, with modified chemistry supporting a surface coating of DOTAP:Chol liposomes and containing the IL-10 transgene, results in functional suppression of pain-related behavior in rats for extended periods. This study is the first demonstration that protocell vectors offer amenable and enduring in vivo biological characteristics that can be applied to spinal gene delivery.
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein (GFAP) Antibody, serum, Chemicon®
Avanti
18:1 TAP (DOTAP), Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand
Avanti
18:1 (Δ9-Cis) PC (DOPC), Avanti Research - A Croda Brand
Avanti
18:1 TAP (DOTAP), Avanti Research - A Croda Brand