MilliporeSigma
Apolipoprotein A-IV is a novel substrate for matrix metalloproteinases.
Journal of biochemistry (2011-12-16)
Ji Yoon Park, Jun Hyoung Park, Wookju Jang, In-Kwan Hwang, In Ja Kim, Hwa-Jung Kim, Kyung-Hyun Cho, Seung-Taek Lee
ABSTRACT
Screening of matrix metalloproteinase (MMP)-14 substrates in human plasma using a proteomics approach previously identified apolipoprotein A-IV (apoA-IV) as a novel substrate for MMP-14. Here, we show that among the tested MMPs, purified apoA-IV is most susceptible to cleavage by MMP-7, and that apoA-IV in plasma can be cleaved more efficiently by MMP-7 than MMP-14. Purified recombinant apoA-IV (44-kDa) was cleaved by MMP-7 into several fragments of 41, 32, 29, 27, 24, 22 and 19 kDa. N-terminal sequencing of the fragments identified two internal cleavage sites for MMP-7 in the apoA-IV sequence, between Glu(185) and Leu(186), and between Glu(262) and Leu(263). The cleavage of lipid-bound apoA-IV by MMP-7 was less efficient than that of lipid-free apoA-IV. Further, MMP-7-mediated cleavage of apoA-IV resulted in a rapid loss of its intrinsic anti-oxidant activity. Based on the fact that apoA-IV plays important roles in lipid metabolism and possesses anti-oxidant activity, we suggest that cleavage of lipid-free apoA-IV by MMP-7 has pathological implications in the development of hyperlipidemia and atherosclerosis.
Sigma-Aldrich
GM6001 MMP Inhibitor, The GM6001 MMP Inhibitor controls the biological activity of MMP. This small molecule/inhibitor is primarily used for Biochemicals applications.