MilliporeSigma
The base excision repair pathway.
Trends in biochemical sciences (1995-10-01)
E Seeberg, L Eide, M Bjørås
ABSTRACT
The base excision repair pathway has evolved to protect cells from the deleterious effects of endogenous DNA damage induced by hydrolysis, reactive oxygen species and other intracellular metabolites that modify DNA base structure. However, base excision repair is also important to resist lesions produced by ionizing radiation and strong alkylating agents, which are similar to those induced by endogenous factors.
Sigma-Aldrich
Fpg Protein from Escherichia coli, ≥90% (SDS-PAGE), buffered aqueous glycerol solution, >20,000 units/mg protein, suitable for genomic analysis