MilliporeSigma
  • Lethal myelofibrosis induced by Bmi1-deficient hematopoietic cells unveils a tumor suppressor function of the polycomb group genes.
Lethal myelofibrosis induced by Bmi1-deficient hematopoietic cells unveils a tumor suppressor function of the polycomb group genes.
The Journal of experimental medicine (2012-02-22)
Hideyuki Oguro, Jin Yuan, Satomi Tanaka, Satoru Miyagi, Makiko Mochizuki-Kashio, Hitoshi Ichikawa, Satoshi Yamazaki, Haruhiko Koseki, Hiromitsu Nakauchi, Atsushi Iwama
ABSTRACT
Polycomb-group (PcG) proteins form the multiprotein polycomb repressive complexes (PRC) 1 and 2, and function as transcriptional repressors through histone modifications. They maintain the proliferative capacity of hematopoietic stem and progenitor cells by repressing the transcription of tumor suppressor genes, namely Ink4a and Arf, and thus have been characterized as oncogenes. However, the identification of inactivating mutations in the PcG gene, EZH2, unveiled a tumor suppressor function in myeloid malignancies, including primary myelofibrosis (PMF). Here, we show that loss of another PcG gene, Bmi1, causes pathological hematopoiesis similar to PMF. In a mouse model, loss of Bmi1 in Ink4a-Arf(-/-) hematopoietic cells induced abnormal megakaryocytopoiesis accompanied by marked extramedullary hematopoiesis, which eventually resulted in lethal myelofibrosis. Absence of Bmi1 caused derepression of a cohort of genes, including Hmga2, which is an oncogene overexpressed in PMF. Chromatin immunoprecipitation assays revealed that Bmi1 directly represses the transcription of Hmga2. Overexpression of Hmga2 in hematopoietic stem cells induced a myeloproliferative state with enhanced megakaryocytopoiesis in mice, implicating Hmga2 in the development of pathological hematopoiesis in the absence of Bmi1. Our findings provide the first genetic evidence of a tumor suppressor function of Bmi1 and uncover the role of PcG proteins in restricting growth by silencing oncogenes.
Sigma-Aldrich
Anti-ubiquityl-Histone H2A Antibody, clone E6C5, clone E6C5, Upstate®, from mouse
Sigma-Aldrich
Rabbit Anti-Mouse IgMµ Antibody, mu chain, Upstate®, from rabbit