MilliporeSigma
  • Poly(L-Lysine)-pyranine-3 coacervate mediated nanoparticle-assembly: fabrication of dynamic pH-responsive containers.
Poly(L-Lysine)-pyranine-3 coacervate mediated nanoparticle-assembly: fabrication of dynamic pH-responsive containers.
Chemical communications (Cambridge, England) (2011-11-30)
Arlin Jose Amali, Shashi Singh, Nandini Rangaraj, Digambara Patra, Rohit Kumar Rana
ABSTRACT
Counter-ion condensation of Poly(L-Lysine) in the presence of pyranine-3 generates spherical coacervates, which then template the assembly of silica nanoparticles to form microcapsule structures that dynamically control the optical ratiometric sensing of both the change in pH and release of the probe molecule.
Sigma-Aldrich
Poly-L-lysine hydrobromide, mol wt 4,000-15,000 by viscosity
Sigma-Aldrich
Poly-L-lysine hydrobromide, mol wt 30,000-70,000
Sigma-Aldrich
Poly-L-lysine hydrobromide, mol wt 15,000-30,000 by viscosity
Sigma-Aldrich
Poly-L-lysine hydrochloride, mol wt >30,000
Sigma-Aldrich
Poly-L-lysine hydrochloride, mol wt 15,000-30,000