MilliporeSigma
  • Integrin receptors play a role in the internalin B-dependent entry of Listeria monocytogenes into host cells.
Integrin receptors play a role in the internalin B-dependent entry of Listeria monocytogenes into host cells.
Cellular & molecular biology letters (2010-06-08)
Clementina Auriemma, Maurizio Viscardi, Simona Tafuri, Luigi Michele Pavone, Federico Capuano, Laura Rinaldi, Rossella Della Morte, Giuseppe Iovane, Norma Staiano
ABSTRACT
Listeria monocytogenes enters non-phagocytic cells by binding its surface proteins inlA (internalin) and inlB to the host's E-cadherin and Met, respectively. The two internalins play either separate or cooperative roles in the colonization of infected tissues. Here, we studied bacterial uptake into HeLa cells using an L. monocytogenes mutant strain (DeltainlA) carrying a deletion in the gene coding for inlA. The DeltainlA mutant strain showed the capability to invade HeLa cells. The monoclonal anti-beta(3)- and anti-beta(1)-integrin subunit antibodies prevented bacterial uptake into the cells, while the anti-beta(2)- and anti-beta(4)-integrin subunit antibodies failed to affect L. monocytogenes entry into HeLa cells. Three structurally distinct disintegrins (kistrin, echistatin and flavoridin) also inhibited bacterial uptake, showing different potencies correlated to their selective affinity for the beta(3)- and beta(1)-integrin subunits. In addition to inducing Met phosphorylation, infection of cells by the L. monocytogenes DeltainlA mutant strain promoted the tyrosine phosphorylation of the focal adhesion-associated proteins FAK and paxillin. Our findings provide the first evidence that beta(3)- and beta(1)-integrin receptors play a role in the inlB-dependent internalization of L. monocytogenes into host cells.
Sigma-Aldrich
Anti-Integrin β3 Antibody, clone B3A, azide free, clone B3A, Chemicon®, from mouse
Sigma-Aldrich
Anti-FAK Antibody, Upstate®, from rabbit