MilliporeSigma
  • Tuftsin derivatives of FITC, Tb-DOTA or Gd-DOTA as potential macrophage-specific imaging biomarkers.
Tuftsin derivatives of FITC, Tb-DOTA or Gd-DOTA as potential macrophage-specific imaging biomarkers.
Contrast media & molecular imaging (2010-08-28)
Jianghua Feng, Marco M Meloni, Stuart M Allan, Stephen Faulkner, Johanna Narvainen, Rishma Vidyasagar, Risto Kauppinen
ABSTRACT
Fluorescein- and terbium-labelled tuftsin (Thr-Lys-Pro-Arg) and pentapeptide (Thr-Lys-Pro-Pro-Arg) were synthesized and their properties were evaluated in vitro by luminescence spectrometry and confocal microscopy as fluorescence probes to target macrophage cells in biological systems. An increase in fluorescence of macrophages incubated with varying concentrations of fluorescein isothiocyanate or Tb-DOTA-tuftsin/pentapeptide conjugates was observed in a concentration-dependent manner. Tb-DOTA-pentapeptide had a greater affinity to macrophages than Tb-DOTA-tuftsin. Lipopolysaccharide (LPS) stimulation strengthened the internalization of peptide conjugates by macrophages through the tuftsin receptor mechanism. Tb-DOTA-tuftsin/pentapeptide conjugates are likely to be a promising optical reagents as probes of the immune response with involvement of macrophage cells in a variety of diseases. Gd-DOTA-tuftsin conjugate was also evaluated as a cell-specific contrast agent in in vitro MRI experiments. In this context, the macrophages labelled by Gd-DOTA-tuftsin were highly magnetic and detectable by MRI, which confirms that this vectorized MRI probe has the potential to image macrophage-mediated inflammation in diseases like brain traumas and stroke. Tuftsin receptor-specific biological-function domain may have a modified in vivo biodistribution profile, bioavailability and pharmacokinetics subsequent to its conjugation to a metal ion-binding backbone.