MilliporeSigma
  • Limitations of electrospray ionization in the analysis of a heterogeneous mixture of naturally occurring hydrophilic and hydrophobic compounds.
Limitations of electrospray ionization in the analysis of a heterogeneous mixture of naturally occurring hydrophilic and hydrophobic compounds.
Rapid communications in mass spectrometry : RCM (2010-10-14)
Antonio Nebbioso, Alessandro Piccolo, Michael Spiteller
ABSTRACT
A model heterogeneous mixture of a hydrophilic tripeptide (phenylalanine-glycine-glycine, PGG) and hydrophobic organic acids ((12)C- and (13)C-octanoic acid and pentadecanoic acid) was subjected to electrospray ionization mass spectrometry (ESI-MS). The objective was to verify the previously noted inconsistencies in ESI-MS of complex environmental samples such as humic materials from either aquatic or terrestrial origins. The hydrophobic organic acids, either alone or together, reduced significantly the ESI-MS detection of the tripeptide molecular and self-associated ions at a concentration that was an order of magnitude lower than that of PGG. The most intense peaks were invariably those of the octanoic acid as either deprotonated, self-associated, or acetate-clustered molecules. The presence of equimolar amounts of PGG and organic acids yielded similar results, but with a significant increased detection of PDA and a smaller depression of the PGG signals. This behaviour is attributed to a different electrospray ionization of the mixture compounds depending on their most probable positioning at the surface of the evaporating droplet. The most favoured positioning of hydrophobic molecules at the aqueous-gas interphase allows preferential evaporation of hydrophobic ions whereas the hydrophilic molecules are retained in the droplet interior, and, their ESI-MS detection depressed. These findings suggest that the electrospray ionization of different molecules present in complex heterogeneous mixtures of environmental significance such as humic substances is limited by their concentration and reciprocal attracting forces.
Sigma-Aldrich
Phe-Gly-Gly
Pricing and availability is not currently available.