MilliporeSigma
L-Phenylalanyl-L-alanine dihydrate.
Acta crystallographica. Section C, Crystal structure communications (2001-05-16)
C H Görbitz
ABSTRACT
A new type of molecular arrangement for dipeptides is observed in the crystal structure of L-phenylalanyl-L-alanine dihydrate, C12H16N2O3-2H2O. Two L-Phe and two L-Ala side chains aggregate into large hydrophobic columns within a three-dimensional hydrogen-bond network.
Sigma-Aldrich
Phe-Ala