MilliporeSigma
  • Intercellular communication via gap junctions in activated rat hepatic stellate cells.
Intercellular communication via gap junctions in activated rat hepatic stellate cells.
Gastroenterology (2005-02-03)
Richard Fischer, Roland Reinehr, Thuy Phung Lu, Alexandra Schönicke, Ulrich Warskulat, Hans Peter Dienes, Dieter Häussinger
ABSTRACT
Gap junctional communication was studied in quiescent and activated hepatic stellate cells. Connexin expression and intercellular dye transfer were studied in rat hepatic stellate cells in culture and in vivo. Protein expression of connexin 43 was up-regulated in activated hepatic stellate cells in vivo and in vitro and was mainly localized on the cell surface, whereas connexin 26 was found intracellularly. In contrast to hepatocytes, hepatic stellate cells do not express connexin 32. Confluent hepatic stellate cells in culture communicate via gap junctions, resulting in lucifer yellow transfer and propagation of intracellular calcium signals. Phorbol ester induces a protein kinase C-dependent hyperphosphorylation and degradation of connexin 43 and inhibits intercellular communication on a short-term time scale. At the long-term level, vitamin D(3) , lipopolysaccharide, thyroid hormone T(3), dexamethasone, platelet-derived growth factor, endothelin 1, and interleukin 1beta up-regulate connexin 43 protein and messenger RNA expression and enhance intercellular communication. Slight down-regulation of connexin 43 is observed in response to vitamin A. Connexin 43 induction by endothelin 1 is inhibited by both endothelin A and endothelin B receptor antagonists. In coculture systems, hepatic stellate cells communicate with each other, which is suggestive of a syncytial organization, but no communication was found between hepatic stellate cells and other liver cell types. As shown by immunohistochemistry and electron microscopy, gap junctions are formed between activated hepatic stellate cells in vivo. Gap junctional communication occurs between hepatic stellate cells, is enhanced after activation, and underlies complex regulation by cytokines, hormones, and vitamins.
Sigma-Aldrich
Anti-Connexin 45 Antibody, near CT, cytoplasmic, clone 5B9.2, clone 5B9.2, Chemicon®, from mouse