MilliporeSigma
  • Design and synthesis of a kininogen-based selective inhibitor of thrombin-induced platelet aggregation.
Design and synthesis of a kininogen-based selective inhibitor of thrombin-induced platelet aggregation.
Peptide research (1994-01-01)
R Matsueda, H Umeyama, R N Puri, H N Bradford, R W Colman
ABSTRACT
Thrombin-induced platelet aggregation has been suggested to play an important role in reocclusion following thrombolytic therapy or angioplasty for treatment of myocardial infarction. We previously demonstrated that thrombin-induced platelet aggregation is indirectly mediated by intracellularly activated calpain expressed on the platelet surface through the cleavage of aggregin, a putative ADP-receptor, and that high molecular weight kininogen (HK), a naturally occurring thiol protease inhibitor, modulates thrombin-induced platelet aggregation. Considering the substrate specificity of calpain and the conserved sequence in HK, we studied selective inhibitors of thrombin-induced platelet aggregation by the affinity labeling approach with an S-3-nitro-2-pyridinesulfenyl (Npys) group. H-Phe-Gln-Val-Val-Cys (Npys)-Gly-NH2, which combines chemical and structural features of calpain substrate specificity and the conserved sequence in HK, selectively inhibited thrombin-induced platelet aggregation. It did not inhibit the aggregatory effects of other platelet agonists, and did not inhibit amidolytic activity of thrombin and thrombin-induced platelet shape change. The design and synthesis of such inhibitors could lead to the development of a new class of inhibitors that selectively block thrombin-induced platelet aggregation.
Sigma-Aldrich
3-Nitro-2-pyridinesulfenyl chloride, 95%