MilliporeSigma
Recombinant adenovirus vectors for cytokine gene therapy in mice.
The Journal of allergy and clinical immunology (1999-05-18)
H Kurata, C B Liu, J Valkova, A E Koch, H Yssel, Y Hirabayashi, T Inoue, T Yokota, K Arai
ABSTRACT
Adenoviruses have several specific features useful for gene therapy. They infect various lineages of cells irrespective of cell cycle status. However, the exact mechanism of their infection and in vivo kinetics as a gene expression vector have not been elucidated. Using adenovirus vectors expressing marker genes, we examined the infectivity of these vectors (including cellular and tissue tropism), the duration and intensity of transgene expression, and the side effects. Various cells were infected with adenovirus expressing LacZ gene at various doses, and beta-galactosidase activity was measured and compared in relation with dose, time course, and cellular vitronectin receptor. Mice were injected with adenoviruses expressing LacZ, luciferase and GM-CSF, and in vivo gene expression was examined. Adenovirus infection induced viral dose-dependent transgene expression that persisted for 2 weeks. Adherent cells were infected much more efficiently than nonadherent cells, probably because the former expressed much higher levels of the vitronectin receptor, one of the main receptors for adenovirus. Studies performed in mice with luciferase-expressing adenovirus revealed that the liver was the main target organ after intravenous injection and showed that the intravenous route was superior to other routes with regard to transgene expression. After intravenous injection of adenovirus expressing human GM-CSF, there was a transient and dose-dependent increase in the serum level of this cytokine. Administration of adenovirus expressing mouse GM-CSF enhanced hematopoiesis in the spleen and bone marrow. These results indicated that adenoviruses can be used for in vivo cytokine gene therapy but suggested the necessity of taking into consideration the route of administration, the duration of transgene expression, the toxic dose, and host immune reactions.
Sigma-Aldrich
Anti-Laminin-1 A&B chains Antibody, cross region, clone AL-2, clone AL-2, Chemicon®, from rat