MilliporeSigma
  • Synthesis of potent antagonists of substance P by modifying the methionyl and glutaminyl residues of its C-terminal hexapeptide and without using D-amino acids.
Synthesis of potent antagonists of substance P by modifying the methionyl and glutaminyl residues of its C-terminal hexapeptide and without using D-amino acids.
International journal of peptide and protein research (1993-04-01)
A Manolopoulou, K Karagiannis, G Stavropoulos, C Poulos, C C Jordan, R M Hagan
ABSTRACT
Analogues of [Orn6]-SP6-11 have been synthesized in which the Met11-NH2 residue is replaced by the alpha, gamma-dimethyl, alpha, gamma-dibenzyl and alpha, gamma-di-tert-butyl esters of glutamic acid. These analogues were tested in three in vitro preparations representative of NK-1, NK-2 and NK-3 receptor types for agonist and antagonist activity. The dimethyl analogue is a selective full agonist in the NK-1 receptor type and a weak antagonist in the other two receptor types, while the dibenzyl and the di-tert-butyl analogues are potent antagonists in the NK-1 receptor type and weak antagonists in the other two receptor types. It is concluded that appropriate modification at the alpha-carboxamide and the side chain of the methionine residue of substance P may induce antagonism without using D-amino acids.
Sigma-Aldrich
L-Glutamic acid di-tert-butyl ester hydrochloride