MilliporeSigma
  • Sequence and functional analysis of the cloned Neisseria meningitidis CMP-NeuNAc synthetase.
Sequence and functional analysis of the cloned Neisseria meningitidis CMP-NeuNAc synthetase.
FEMS microbiology letters (1992-09-15)
U Edwards, M Frosch
ABSTRACT
The CMP-N-acetylneuraminic acid (CMP-NeuNAc) synthetase gene of Neisseria meningitidis group B is located on a 2.3-kb EcoRI fragment within the cps gene cluster. Nucleotide sequence determination of the gene encoding the CMP-NeuNAc synthetase revealed a 515-bp open reading frame that can encode a 18.9-kDA protein. A computer data base scan revealed a 59.4% identity to the CMP-NeuNAc synthetase gene of E. coli K1. Enzymatic activity was confirmed in vitro and in vivo. Transformation of the CMP-NeuNAc defective E. coli K1 strain EV5 with the meningococcal CMP-NeuNAc synthetase could complement the defect in E. coli.
Sigma-Aldrich
Cytidine 5′-triphosphate disodium salt hydrate, ≥90.0% (HPLC)