MilliporeSigma
  • Monoclonal antibodies capable of distinguishing epizootic from enzootic varieties of subtype 1 Venezuelan equine encephalitis viruses in a rapid indirect immunofluorescence assay.
Monoclonal antibodies capable of distinguishing epizootic from enzootic varieties of subtype 1 Venezuelan equine encephalitis viruses in a rapid indirect immunofluorescence assay.
Journal of clinical microbiology (1997-07-01)
J T Roehrig, R A Bolin
ABSTRACT
We used previously characterized murine monoclonal antibodies to develop a panel useful in subtyping Venezuelan equine encephalitis (VEE) viruses by an indirect fluorescent antibody assay. This panel worked well with either prototype VEE viruses or a series of more recent VEE virus isolates. The panel is particularly useful for rapidly differentiating VEE viruses with epidemic-epizootic potential from other endemic varieties of this virus. Using this panel, we identified an antigenic variant of prototype VEE subtype 1E virus currently present in Mexico. This antigenic change in the E2 glycoprotein was confirmed by enzyme-linked immunosorbent assay. Because VEE virus virulence has been associated in part with the E2 glycoprotein, this observed antigenic change in the 1E virus E2 glycoprotein may explain the apparent equine virulence of this unusual VEE 1E virus.
Sigma-Aldrich
Anti-St. Louis Encephalitis Antibody, MSI-7, clone 6B6C-1, clone 6b6c-1, Chemicon®, from mouse