MilliporeSigma
  • Analysis of methadone and metabolites in biological fluids with gas chromatography--mass spectrometry.
Analysis of methadone and metabolites in biological fluids with gas chromatography--mass spectrometry.
Journal of chromatography (1982-09-10)
G I Kang, F S Abbott
ABSTRACT
The analysis of methadone and its metabolites in biological fluids by gas chromatography--mass spectrometry is described with deuterated methadone and metabolites as internal standards. The method allowed the determination of 20 ng methadone in 0.5 ml of plasma or saliva. Mean saliva to plasma ratio of methadone for two patients was determined to be 0.51 +/- 0.13. Methadone and 2-ethylidene-1,5-dimethyl-3,3-diphenylpyrrolidine (EDDP) in urine were measured by selected ion monitoring. Gas chromatography--mass spectrometry was found to have advantages over conventional gas chromatographic methods in terms of ratio analysis. 1,5-Dimethyl-3,3-diphenyl-2-pyrrolidone previously reported as a metabolite was shown to result primarily from the decomposition of EDDP free base.