MilliporeSigma
Interaction of convulsive ligands with benzodiazepine receptors.
Science (New York, N.Y.) (1982-06-11)
C Braestrup, R Schmiechen, G Neef, M Nielsen, E N Petersen
ABSTRACT
The gamma-aminobutyric acid (GABA)-benzodiazepine receptor complex, which is composed of distinct proteins embedded in the neuronal plasma membrane, is important for several effects of benzodiazepines, including protection afforded against convulsions. During structural modification of ethyl beta-carboline-3-carboxylate an agent was discovered which has high affinity for brain benzodiazepine receptors but which is a potent convulsant. Also in contrast to benzodiazepines, this type of benzodiazepine receptor ligand favors benzodiazepine receptors in the non-GABA-stimulated conformation, which may explain the convulsive properties.