MilliporeSigma
  • Unbiased statistical analysis for multi-stage proteomic search strategies.
Unbiased statistical analysis for multi-stage proteomic search strategies.
Journal of proteome research (2009-12-02)
Logan J Everett, Charlene Bierl, Stephen R Master
ABSTRACT
"Multi-stage" search strategies have become widely accepted for peptide identification and are implemented in a number of available software packages. We describe limitations of these strategies for validation and decoy-based statistical analyses and demonstrate these limitations using a set of control sample spectra. We propose a solution that corrects the statistical deficiencies and describe its implementation using the open-source software X!Tandem.
Sigma-Aldrich
Trypsin Spin Columns