MilliporeSigma
  • Targeting of DC-SIGN on human dendritic cells by minor fimbriated Porphyromonas gingivalis strains elicits a distinct effector T cell response.
Targeting of DC-SIGN on human dendritic cells by minor fimbriated Porphyromonas gingivalis strains elicits a distinct effector T cell response.
Journal of immunology (Baltimore, Md. : 1950) (2009-10-16)
Amir E Zeituni, Ravi Jotwani, Julio Carrion, Christopher W Cutler
ABSTRACT
The oral mucosal pathogen Porphyromonas gingivalis expresses at least two adhesins: the 67-kDa mfa-1 (minor) fimbriae and the 41-kDa fimA (major) fimbriae. In periodontal disease, P. gingivalis associates in situ with dermal dendritic cells (DCs), many of which express DC-SIGN (DC-specific ICAM-3 grabbing nonintegrin; CD209). The cellular receptors present on DCs that are involved in the uptake of minor/major fimbriated P. gingivalis, along with the effector immune response induced, are presently unclear. In this study, stably transfected human DC-SIGN(+/-) Raji cell lines and monocyte-derived DCs (MoDCs) were pulsed with whole, live, wild-type Pg381 or isogenic major (DPG-3)-, minor (MFI)-, or double fimbriae (MFB)-deficient mutant P. gingivalis strains. The influence of blocking Abs, carbohydrates, full-length glycosylated HIV-1 gp120 envelope protein, and cytochalasin D on the uptake of strains and on the immune responses was determined in vitro. We show that the binding of minor fimbriated P. gingivalis strains to Raji cells and MoDCs is dependent on DC-SIGN, whereas the double fimbriae mutant strain does not bind. Binding to DC-SIGN on MoDCs is followed by the internalization of P. gingivalis into DC-SIGN-rich intracellular compartments, and MoDCs secrete low levels of inflammatory cytokines and remain relatively immature. Blocking DC-SIGN with HIV-1 gp120 prevents the uptake of minor fimbriated strains and deregulates the expression of inflammatory cytokines. Moreover, MoDCs promote a Th2 or Th1 effector response, depending on whether they are pulsed with minor or major fimbriated P. gingivalis strains, respectively, suggesting distinct immunomodulatory roles for the two adhesins of P. gingivalis.
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone P4C10, azide free, clone P4C10, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin β2 [CD18] Antibody, clone MEM 48, Chemicon®, from mouse