MilliporeSigma
  • Toxin-binding proteins isolated from yellow mealworm Tenebrio molitor and wax moth Galleria mellonella.
Toxin-binding proteins isolated from yellow mealworm Tenebrio molitor and wax moth Galleria mellonella.
Biochemistry. Biokhimiia (2011-05-17)
N V Bulushova, D P Zhuzhikov, L I Lyutikova, N E Kirillova, I A Zalunin, G G Chestukhina
ABSTRACT
A 67-kDa protein that can specifically bind the activated Cry9A endotoxin under ligand-blotting conditions was purified from midgut epithelium apical membranes of wax moth Galleria mellonella by affinity chromatography. N-Terminal amino acid sequencing enabled identification of this protein as aminopeptidase N. In similar experiments, 66- and 58-kDa proteins specific to endotoxin Cry3A were isolated from the midgut epithelium apical membranes of Tenebrio molitor larvae. Mass spectrometry showed close similarity of the 58-kDa protein to the Tenebrio molitor α-amylase.
Sigma-Aldrich
ω-Aminohexyl–Agarose, saline suspension