MilliporeSigma
  • Coulometric D-fructose biosensor based on direct electron transfer using D-fructose dehydrogenase.
Coulometric D-fructose biosensor based on direct electron transfer using D-fructose dehydrogenase.
Analytical chemistry (2009-11-17)
Seiya Tsujimura, Akiko Nishina, Yuji Kamitaka, Kenji Kano
ABSTRACT
This paper describes a batch-type coulometric d-fructose biosensor based on direct electron transfer reaction of d-fructose dehydrogenase (FDH) adsorbed on a porous carbon electrode surface. The adsorbed-FDH electrodes catalyzed the electrochemical two-electron oxidation of d-fructose to 5-keto-d-fructose without a mediator. Nanostructured carbon particle-modified electrodes were used for the coulometric d-fructose biosensor to enhance the catalytic current density. The electric charge for the d-fructose oxidation gained by the biocoulometric measurement was in good agreement with the theoretical value corresponding to d-fructose amount in the range from 1 to 100 mM with a sample volume of 1 muL. This method is also applicable to the determination of several oligo/polysaccharides containing the d-fructose unit, in combination with specific hydrolases to yield d-fructose. An example was demonstrated by sucrose determination in which the electrode modified with FDH and invertase was used as a working electrode. To address the problem of electroactive interferences such as ascorbate, the electric charge at the FDH-free electrode was subtracted from the total charge obtained at the FDH-adsorbed electrode. The d-fructose concentrations in several beverages were successfully determined with this method.