MilliporeSigma
  • Transactivation of platelet-derived growth factor receptor alpha by the GTPase-deficient activated mutant of Galpha12.
Transactivation of platelet-derived growth factor receptor alpha by the GTPase-deficient activated mutant of Galpha12.
Molecular and cellular biology (2005-12-16)
Rashmi N Kumar, Ji Hee Ha, Rangasudhagar Radhakrishnan, Danny N Dhanasekaran
ABSTRACT
The GTPase-deficient, activated mutant of Galpha12 (Galpha12Q229L, or Galpha12QL) induces neoplastic growth and oncogenic transformation of NIH 3T3 cells. Using microarray analysis, we have previously identified a role for platelet-derived growth factor receptor alpha (PDGFRalpha) in Galpha12-mediated cell growth (R. N. Kumar et al., Cell Biochem. Biophys. 41:63-73, 2004). In the present study, we report that Galpha12QL stimulates the functional expression of PDGFRalpha and demonstrate that the expression of PDGFRalpha by Galpha12QL is dependent on the small GTPase Rho. Our results indicate that it is cell type independent as the transient expression of Galpha12QL or the activation of Galpha12-coupled receptors stimulates the expression of PDGFRalpha in NIH 3T3 as well as in human astrocytoma 1321N1 cells. Furthermore, we demonstrate the presence of an autocrine loop involving PDGF-A and PDGFRalpha in Galpha12QL-transformed cells. Analysis of the functional consequences of the Galpha12-PDGFRalpha signaling axis indicates that Galpha12 stimulates the phosphatidylinositol 3-kinase (PI3K)-AKT signaling pathway through PDGFR. In addition, we show that Galpha12QL stimulates the phosphorylation of forkhead transcription factor FKHRL1 via AKT in a PDGFRalpha- and PI3K-dependent manner. Since AKT promotes cell growth by blocking the transcription of antiproliferative genes through the inhibitory phosphorylation of forkhead transcription factors, our results describe for the first time a PDGFRalpha-dependent signaling pathway involving PI3K-AKT-FKHRL1, regulated by Galpha12QL in promoting cell growth. Consistent with this view, we demonstrate that the expression of a dominant negative mutant of PDGFRalpha attenuated Galpha12-mediated neoplastic transformation of NIH 3T3 cells.
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse